Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Hydroxymethyl-4-methoxybenzeneboronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
908142-03-0
C8H11BO4
181.982
MFCD14155851
DOQUCBSZQOPLGT-UHFFFAOYSA-N
3-hydroxymethyl-4-methoxyphenyl boronic acid,3-hydroxymethyl-4-methoxyphenylboronic acid,3-hydroxymethyl-4-methoxyphenylboronic acid,3-hydroxymethyl-4-methoxyphenyl boronic acid,ksc005q4b,3-hydroxymethyl-4-methoxybenzeneboronic acid,3-hydroxymethyl-4-methoxyphenyl boronicacid,3-hydroxymethyl-4-methoxybenzeneboronic acid,b-3-hydroxymethyl-4-methoxyphenyl-boronic acid,boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
53407782
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
B(C1=CC(=C(C=C1)OC)CO)(O)O
908142-03-0
C8H11BO4
181.982
MFCD14155851
DOQUCBSZQOPLGT-UHFFFAOYSA-N
3-hydroxymethyl-4-methoxyphenyl boronic acid,3-hydroxymethyl-4-methoxyphenylboronic acid,3-hydroxymethyl-4-methoxyphenylboronic acid,3-hydroxymethyl-4-methoxyphenyl boronic acid,ksc005q4b,3-hydroxymethyl-4-methoxybenzeneboronic acid,3-hydroxymethyl-4-methoxyphenyl boronicacid,3-hydroxymethyl-4-methoxybenzeneboronic acid,b-3-hydroxymethyl-4-methoxyphenyl-boronic acid,boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
53407782
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
B(C1=CC(=C(C=C1)OC)CO)(O)O
Applications
It is an important intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 1g
Color White
Beilstein 15501236
Formula Weight 181.98
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Hydroxymethyl-4-methoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning