Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Hydroxymethyl)pyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
100-55-0
C6H7NO
109.13
MFCD00006407
MVQVNTPHUGQQHK-UHFFFAOYSA-N
3-pyridinemethanol,nicotinyl alcohol,3-hydroxymethyl pyridine,3-pyridylmethanol,nicotinic alcohol,3-pyridylcarbinol,roniacol,pyridylcarbinol,3-pyridinylmethanol,pyridine-3-carbinol
7510
pyridin-3-ylmethanol
C1=CC(=CN=C1)CO
100-55-0
C6H7NO
109.13
MFCD00006407
MVQVNTPHUGQQHK-UHFFFAOYSA-N
3-pyridinemethanol,nicotinyl alcohol,3-hydroxymethyl pyridine,3-pyridylmethanol,nicotinic alcohol,3-pyridylcarbinol,roniacol,pyridylcarbinol,3-pyridinylmethanol,pyridine-3-carbinol
7510
pyridin-3-ylmethanol
C1=CC(=CN=C1)CO
Specific Gravity 1.124
Flash Point 149°C
Melting Point -7°C
Boiling Point 259°C
Refractive Index 1.5425 to 1.5455
Density 1.1240g/mL
Quantity 500g
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)CH2OH
Beilstein 21, V,2, 172
Merck Index 15, 6612
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in ether
Formula Weight 109.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-(Hydroxymethyl)pyridine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-864-6