Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypiperidine, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
6859-99-0
C5H11NO
101.15
MFCD00014591
BIWOSRSKDCZIFM-UHFFFAOYSA-N
3-hydroxypiperidine,3-piperidinol,3-hydroxypiperadine,3-hydroxy piperidine,+/--3-hydroxypiperidine,racemic-3r-piperidin-3-ol,3-s-hydroxypiperidine,piperidine-3-ol,ppiperidin-3-ol,3-hydroxy-piperdine
23293
piperidin-3-ol
C1CC(CNC1)O
6859-99-0
C5H11NO
101.15
MFCD00014591
BIWOSRSKDCZIFM-UHFFFAOYSA-N
3-hydroxypiperidine,3-piperidinol,3-hydroxypiperadine,3-hydroxy piperidine,+/--3-hydroxypiperidine,racemic-3r-piperidin-3-ol,3-s-hydroxypiperidine,piperidine-3-ol,ppiperidin-3-ol,3-hydroxy-piperdine
23293
piperidin-3-ol
C1CC(CNC1)O
Flash Point >100°C
Boiling Point 67°C to 69°C (2.0mmHg)
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
Quantity 50g
Melting Point 56°C to 62°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 21, V,1, 28
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform
Formula Weight 101.15
Physical Form Crystalline Powder and Lumps
Percent Purity 98%
Chemical Name or Material 3-Hydroxypiperidine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 229-957-4

Recommended Storage : Refrigerator +4°C