Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Hydroxypiperidine hydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 196.6°C to 198.7°C |
Color | White |
Elemental Analysis | 42.97 to 44.28% C, 9.90 to 10.33% N |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Quantity | 5 g |
Formula Weight | 137.61 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing. |
EINECSNumber | 264-644-6 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction