Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypiperidine hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
64051-79-2
C5H11NO·ClH
137.61
VLECDMDGMKPUSK-UHFFFAOYSA-N
3-hydroxypiperidine hydrochloride,piperidin-3-ol hydrochloride,3-hydroxypiperidine hcl,3-piperidinol hydrochloride,piperidine-3-ol hydrochloride,3-hydroxy-piperidine hydrochloride,3-piperidinol, hydrochloride,piperidine-3-ol hcl,3-hydroxypiperidine.hcl,3-hydroxypiperidinehydrochloride
2723962
piperidin-3-ol;hydrochloride
C1CC(CNC1)O.Cl
64051-79-2
C5H11NO·ClH
137.61
VLECDMDGMKPUSK-UHFFFAOYSA-N
3-hydroxypiperidine hydrochloride,piperidin-3-ol hydrochloride,3-hydroxypiperidine hcl,3-piperidinol hydrochloride,piperidine-3-ol hydrochloride,3-hydroxy-piperidine hydrochloride,3-piperidinol, hydrochloride,piperidine-3-ol hcl,3-hydroxypiperidine.hcl,3-hydroxypiperidinehydrochloride
2723962
piperidin-3-ol;hydrochloride
C1CC(CNC1)O.Cl
Color White
Elemental Analysis 42.97 to 44.28% C, 9.90 to 10.33% N
Infrared Spectrum Authentic
Quantity 100g
Melting Point 196.6°C to 198.7°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 137.61
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Hydroxypiperidine hydrochloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 264-644-6