Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypropionitrile 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
109-78-4
C3H5NO
71.08
MFCD00002826
WSGYTJNNHPZFKR-UHFFFAOYSA-N
3-hydroxypropionitrile,hydracrylonitrile,2-cyanoethanol,ethylene cyanohydrin,propanenitrile, 3-hydroxy,beta-cyanoethanol,glycol cyanohydrin,methanolacetonitrile,hydroxypropanenitrile,2-hydroxycyanoethane
8011
3-hydroxypropanenitrile
C(CO)C#N
109-78-4
C3H5NO
71.08
MFCD00002826
WSGYTJNNHPZFKR-UHFFFAOYSA-N
3-hydroxypropionitrile,hydracrylonitrile,2-cyanoethanol,ethylene cyanohydrin,propanenitrile, 3-hydroxy,beta-cyanoethanol,glycol cyanohydrin,methanolacetonitrile,hydroxypropanenitrile,2-hydroxycyanoethane
8011
3-hydroxypropanenitrile
C(CO)C#N
Melting Point -46°C
Flash Point 128°C
Boiling Point 228°C
Viscosity <10 mPa.s (20°C)
Specific Gravity 1.04
Density 1.0400g/mL
Refractive Index 1.4246 to 1.4266
Color Yellow
Quantity 1kg
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOCH2CH2CN
Beilstein 03, 298
Merck Index 15, 3848
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,methyl ethyl ketone and,ethyl alcohol,slightly soluble in ether,insoluble in benzene,carbon disulfide and ccl4
Formula Weight 71.08
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3-Hydroxypropionitrile, 98%
Show More Show Less

EINECSNumber : 203-704-8

RTECSNumber : MU5250000

TSCA : TSCA