Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypyridine 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
109-00-2
C5H5NO
95.1
MFCD00006378
GRFNBEZIAWKNCO-UHFFFAOYSA-N
3-hydroxypyridine,3-pyridinol,3-pyridol,3-pyridone,3-oxopyridine,beta-hydroxypyridine,m-hydroxypyridine,3-pyridyl alcohol,3-hydoxypyridine,unii-4kbe4p5b6s
7971
pyridin-3-ol
C1=CC(=CN=C1)O
109-00-2
C5H5NO
95.1
MFCD00006378
GRFNBEZIAWKNCO-UHFFFAOYSA-N
3-hydroxypyridine,3-pyridinol,3-pyridol,3-pyridone,3-oxopyridine,beta-hydroxypyridine,m-hydroxypyridine,3-pyridyl alcohol,3-hydoxypyridine,unii-4kbe4p5b6s
7971
pyridin-3-ol
C1=CC(=CN=C1)O
Infrared Spectrum Authentic
Water 0.2% max.
Packaging Plastic bottle
Quantity 500g
Melting Point 123.0°C to 130.0°C
Color Beige to Brown
Boiling Point 180.0°C (22.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, 46
Fieser 01,486
Solubility Information Solubility in water: 33g/L water (20°C).
Formula Weight 95.1
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3-Hydroxypyridine, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-637-4

RTECSNumber : UU7701400

TSCA : TSCA