Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypyridine-N-oxide 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6602-28-4
C5H5NO2
111.1
MFCD00006200
YMEZKRMAPQIBQH-UHFFFAOYSA-N
3-hydroxypyridine-n-oxide,3-hydroxypyridine 1-oxide,3-hydroxypyridine n-oxide,3-pyridinol n-oxide,pyridin-3-ol 1-oxide,3-pyridinol, 1-oxide,3-hydroxypyridin-1-ium-1-olate,3-pyridinol-1-oxide,3-hydroxypyridine-1-oxide,3-pyridinol 1-oxide
23069
1-oxidopyridin-1-ium-3-ol
C1=CC(=C[N+](=C1)[O-])O
6602-28-4
C5H5NO2
111.1
MFCD00006200
YMEZKRMAPQIBQH-UHFFFAOYSA-N
3-hydroxypyridine-n-oxide,3-hydroxypyridine 1-oxide,3-hydroxypyridine n-oxide,3-pyridinol n-oxide,pyridin-3-ol 1-oxide,3-pyridinol, 1-oxide,3-hydroxypyridin-1-ium-1-olate,3-pyridinol-1-oxide,3-hydroxypyridine-1-oxide,3-pyridinol 1-oxide
23069
1-oxidopyridin-1-ium-3-ol
C1=CC(=C[N+](=C1)[O-])O
Infrared Spectrum Authentic
Melting Point 189°C to 192°C
Quantity 5g
Packaging Glass bottle
Color Yellow to Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 111.1
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 3-Hydroxypyridine-N-oxide, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 229-545-4

RTECSNumber : UU7719100

TSCA : TSCA