Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-3-Hydroxypyrrolidine hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
86070-82-8
C4H10ClNO
123.58
MFCD03001754
QPMSJEFZULFYTB-UHFFFAOYSA-N
pyrrolidin-3-ol hydrochloride,3-hydroxypyrrolidine hydrochloride,3-pyrrolidinol hydrochloride,3-hydroxypyrrolidine hcl,3-pyrrolidinol hcl,3-pyrrolidinol, hydrochloride,3-hydroxypyrrolidinehydrochloride,s-3-hydroxypyrrolidine hcl,3-pyrrolidinol hci,hydroxypyrrolidine hcl
2769408
pyrrolidin-3-ol;hydrochloride
C1CNCC1O.Cl
86070-82-8
C4H10ClNO
123.58
MFCD03001754
QPMSJEFZULFYTB-UHFFFAOYSA-N
pyrrolidin-3-ol hydrochloride,3-hydroxypyrrolidine hydrochloride,3-pyrrolidinol hydrochloride,3-hydroxypyrrolidine hcl,3-pyrrolidinol hcl,3-pyrrolidinol, hydrochloride,3-hydroxypyrrolidinehydrochloride,s-3-hydroxypyrrolidine hcl,3-pyrrolidinol hci,hydroxypyrrolidine hcl
2769408
pyrrolidin-3-ol;hydrochloride
C1CNCC1O.Cl
Quantity 5g
Beilstein 7029051
Formula Weight 123.58
Physical Form Solid
Percent Purity 98%
Chemical Name or Material (±)-3-Hydroxypyrrolidine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning