Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxytyramine Hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
62-31-7
C8H11NO2·HCl
189.64
MFCD00012898
CTENFNNZBMHDDG-UHFFFAOYSA-N
dopamine hydrochloride,3-hydroxytyramine hydrochloride,dopamine hcl,4-2-aminoethyl benzene-1,2-diol hydrochloride,intropin,dopastat,revivan,dynatra,3,4-dihydroxyphenethylamine hydrochloride,cardiosteril
65340
4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
C1=CC(=C(C=C1CCN)O)O.Cl
62-31-7
C8H11NO2·HCl
189.64
MFCD00012898
CTENFNNZBMHDDG-UHFFFAOYSA-N
dopamine hydrochloride,3-hydroxytyramine hydrochloride,dopamine hcl,4-2-aminoethyl benzene-1,2-diol hydrochloride,intropin,dopastat,revivan,dynatra,3,4-dihydroxyphenethylamine hydrochloride,cardiosteril
65340
4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
C1=CC(=C(C=C1CCN)O)O.Cl
Melting Point 241°C to 243°C
Packaging Glass bottle
Sulfated Ash 0.1% max.
Loss on Drying 0.5% max. (105°C, 2 hrs)
Quantity 100g
Color White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (HO)2C6H3CH2CH2NH2·HCl
Beilstein 13, I, 325
Merck Index 15, 3470
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol and hot 95% ethanol,practically insoluble in ether,chloroform,,benzene and toluene
Formula Weight 189.64
Physical Form Crystalline Powder
Percent Purity ≥98.5% (on dried substance)
Chemical Name or Material 3-Hydroxytyramine hydrochloride
Show More Show Less