Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indolepropionic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
830-96-6
C11H11NO2
189.21
MFCD00005660
GOLXRNDWAUTYKT-UHFFFAOYSA-N
3-indolepropionic acid,indole-3-propionic acid,3-1h-indol-3-yl propanoic acid,1h-indole-3-propanoic acid,indolepropionic acid,indolylpropionic acid,3-3-indolyl propionic acid,3-3-indolyl propanoic acid,ipa auxin,1h-indole-3-propionic acid
3744
CHEBI:43580
3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(=CN2)CCC(=O)O
830-96-6
C11H11NO2
189.21
MFCD00005660
GOLXRNDWAUTYKT-UHFFFAOYSA-N
3-indolepropionic acid,indole-3-propionic acid,3-1h-indol-3-yl propanoic acid,1h-indole-3-propanoic acid,indolepropionic acid,indolylpropionic acid,3-3-indolyl propionic acid,3-3-indolyl propanoic acid,ipa auxin,1h-indole-3-propionic acid
3744
CHEBI:43580
3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(=CN2)CCC(=O)O
Water 0.5% Max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 130°C to 136°C
Quantity 50g
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Beilstein 22, 69
Solubility Information Solubility in water: slightly soluble
Formula Weight 189.21
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3-Indolepropionic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 212-600-1