Alfa Aesar™ 3-Iodo-1-phenylpropane, 97%

Manufacturer: Alfa Aesar™ H5342014

Quantity 25g

 View more versions of this product

Catalog No. AAH5342014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4119-41-9
    Molecular Formula C9H11I
    Linear Formula C6H5(CH2)3I
    Formula Weight 246.09
    MDL Number MFCD00019036
    Synonym 3-Iodopropylbenzene
    Chemical Name or Material 3-Iodo-1-phenylpropane

    Specifications

    Formula Weight 246.09
    Linear Formula C6H5(CH2)3I
    Quantity 25g
    Molecular Formula C9H11I
    MDL Number MFCD00019036
    CAS 4119-41-9
    Synonym 3-Iodopropylbenzene
    Flash Point 110°C (230°F)
    Chemical Name or Material 3-Iodo-1-phenylpropane
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit