Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Iodo-4-methoxybenzaldehyde, 96%

Click to view available options
Quantity:
1g
5g
2314-37-6
C8H7IO2
262.046
MFCD01009500
RVWOHBWQJGLXIJ-UHFFFAOYSA-N
3-iodo-4-methoxy-benzaldehyde,benzaldehyde, 3-iodo-4-methoxy,3-iodo-p-anisaldehyde,4-formyl-2-iodoanisole,acmc-1cpd6,3-iodanyl-4-methoxy-benzaldehyde,3-iodo-4-methoxybenzaldehyde,benzaldehyde,3-iodo-4-methoxy
1494376
3-iodo-4-methoxybenzaldehyde
COC1=C(C=C(C=C1)C=O)I
2314-37-6
C8H7IO2
262.046
MFCD01009500
RVWOHBWQJGLXIJ-UHFFFAOYSA-N
3-iodo-4-methoxy-benzaldehyde,benzaldehyde, 3-iodo-4-methoxy,3-iodo-p-anisaldehyde,4-formyl-2-iodoanisole,acmc-1cpd6,3-iodanyl-4-methoxy-benzaldehyde,3-iodo-4-methoxybenzaldehyde,benzaldehyde,3-iodo-4-methoxy
1494376
3-iodo-4-methoxybenzaldehyde
COC1=C(C=C(C=C1)C=O)I
Sensitivity Air & Light Sensitive
Quantity 5g
Color White to White-Yellow
Melting Point 107°C to 112°C
Formula Weight 262.04
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Iodo-4-methoxybenzaldehyde