Alfa Aesar™ 3-Iodo-4-methylaniline, 98%

Manufacturer: Alfa Aesar™ A1148522

Quantity 100g

 View more versions of this product

Catalog No. AAA1148522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35944-64-0
    Molecular Formula C7H8IN
    Formula Weight 233.05
    MDL Number MFCD00047843
    UN Number UN2811
    Beilstein 2353567
    Synonym 4-Amino-2-iodotoluene; 3-Iodo-p-toluidine
    Chemical Name or Material 3-Iodo-4-methylaniline

    Specifications

    Formula Weight 233.05
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym 4-Amino-2-iodotoluene; 3-Iodo-p-toluidine
    RTECS Number XU7700000
    Molecular Formula C7H8IN
    Beilstein 2353567
    Quantity 100g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    MDL Number MFCD00047843
    CAS 35944-64-0
    UN Number UN2811
    Melting Point 34° to 37°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 3-Iodo-4-methylaniline
    Assay 98%

    Description

    Applications
    3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

    Solubility
    Insoluble in water.

    Notes
    Light Sensitive .Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit