Alfa Aesar™ 3-Iodo-4-methylbenzoic acid, 98%

Manufacturer: Alfa Aesar™ A1419706

Quantity 5g

 View more versions of this product

Catalog No. AAA1419706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 82998-57-0
    Molecular Formula C8H7IO2
    Formula Weight 262.05
    MDL Number MFCD00010392
    Beilstein 2436216
    Synonym 3-Iodo-p-toluic acid
    Chemical Name or Material 3-Iodo-4-methylbenzoic acid

    Specifications

    Formula Weight 262.05
    CAS 82998-57-0
    Beilstein 2436216
    Melting Point 206° to 210°C
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C8H7IO2
    MDL Number MFCD00010392
    Synonym 3-Iodo-p-toluic acid
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Iodo-4-methylbenzoic acid
    Assay 98%

    Description

    Applications
    3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).

    Solubility
    Soluble in methanol.

    Notes
    Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit