Alfa Aesar™ 3-Iodobenzoic acid, 98+%

Manufacturer: Alfa Aesar™ A1151530

Quantity 250g

 View more versions of this product

Catalog No. AAA1151530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 618-51-9
    Molecular Formula C7H5IO2
    Formula Weight 248.02
    MDL Number MFCD00002496
    Beilstein 971088
    Chemical Name or Material 3-Iodobenzoic acid

    Specifications

    Formula Weight 248.02
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Melting Point 186° to 188°C
    Molecular Formula C7H5IO2
    CAS 618-51-9
    Beilstein 971088
    Quantity 250g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00002496
    Chemical Name or Material 3-Iodobenzoic acid
    Assay ≥98%

    Description

    Applications
    3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

    Solubility
    Soluble in Chloroform and Methanol. Insoluble in water.

    Notes
    Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit