Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
69113-59-3
C7H4IN
229.02
MFCD00079762
BGARPMGQRREXLN-UHFFFAOYSA-N
m-iodobenzonitrile,3-iodo-benzonitrile,benzonitrile, 3-iodo,1-cyano-3-iodobenzene,3-iodobenzenecarbonitrile,pubchem3935,3-cyanoiodobenzene,3-iodobenzonitrile,ksc358g7h,acmc-1b930
144341
3-iodobenzonitrile
C1=CC(=CC(=C1)I)C#N
69113-59-3
C7H4IN
229.02
MFCD00079762
BGARPMGQRREXLN-UHFFFAOYSA-N
m-iodobenzonitrile,3-iodo-benzonitrile,benzonitrile, 3-iodo,1-cyano-3-iodobenzene,3-iodobenzenecarbonitrile,pubchem3935,3-cyanoiodobenzene,3-iodobenzonitrile,ksc358g7h,acmc-1b930
144341
3-iodobenzonitrile
C1=CC(=CC(=C1)I)C#N
Flash Point >100°C
Melting Point 39.0°C to 44.0°C
Quantity 50g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 229.02
Percent Purity 97%
Chemical Name or Material 3-Iodobenzonitrile, 97%

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning