Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
57455-06-8
C7H7IO
234.04
MFCD00004635
QGCCNWSXJHGUNL-UHFFFAOYSA-N
3-iodophenyl methanol,3-iodobenzyl alcohol,m-iodobenzyl alcohol,benzenemethanol, 3-iodo,benzyl alcohol, m-iodo,3-iodo-benzyl alcohol,3-iodobenzylalcohol,3-iodo-phenyl-methanol,m-iodo benzylalcohol,3-iodophenyl methan-1-ol
42300
(3-iodophenyl)methanol
C1=CC(=CC(=C1)I)CO
57455-06-8
C7H7IO
234.04
MFCD00004635
QGCCNWSXJHGUNL-UHFFFAOYSA-N
3-iodophenyl methanol,3-iodobenzyl alcohol,m-iodobenzyl alcohol,benzenemethanol, 3-iodo,benzyl alcohol, m-iodo,3-iodo-benzyl alcohol,3-iodobenzylalcohol,3-iodo-phenyl-methanol,m-iodo benzylalcohol,3-iodophenyl methan-1-ol
42300
(3-iodophenyl)methanol
C1=CC(=CC(=C1)I)CO
Color Orange-Red to Yellow
Infrared Spectrum Authentic
Flash Point >110°C
Boiling Point 252°C (711.0mmHg)
Packaging Glass bottle
Refractive Index 1.6340 to 1.636
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula IC6H4CH2OH
Formula Weight 234.04
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Iodobenzyl alcohol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 260-744-9