Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Iodoindole, 96%

Click to view available options
Quantity:
250mg
1g
26340-47-6
C8H6IN
243.047
MFCD07778364
FQVDXLYJTMHMCG-UHFFFAOYSA-N
3-iodoindole,1h-indole, 3-iodo,3-indolyl-iodide,3-iodo-indole,3-jod-1h-indole,fqvdxlyjtmhmcg-uhfffaoysa
11207253
3-iodo-1H-indole
C1=CC=C2C(=C1)C(=CN2)I
26340-47-6
C8H6IN
243.047
MFCD07778364
FQVDXLYJTMHMCG-UHFFFAOYSA-N
3-iodoindole,1h-indole, 3-iodo,3-indolyl-iodide,3-iodo-indole,3-jod-1h-indole,fqvdxlyjtmhmcg-uhfffaoysa
11207253
3-iodo-1H-indole
C1=CC=C2C(=C1)C(=CN2)I
Sensitivity Light Sensitive
Quantity 250mg
Color Brown to Cream
Formula Weight 243.04
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Iodoindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C