Alfa Aesar™ 3-Iodophenyl isocyanate, 97%

Manufacturer: Alfa Aesar™ H3080306

Quantity 5g

 View more versions of this product

Catalog No. AAH3080306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23138-56-9
    Molecular Formula C7H4INO
    Formula Weight 245.02
    MDL Number MFCD00037088
    UN Number UN2206
    Chemical Name or Material 3-Iodophenyl isocyanate

    Specifications

    Formula Weight 245.02
    Flash Point >110°C (230°F)
    Molecular Formula C7H4INO
    MDL Number MFCD00037088
    CAS 23138-56-9
    UN Number UN2206
    Boiling Point 55°C (0.2mmHg)
    Quantity 5g
    Odor Pungent
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H334-H302-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
    Chemical Name or Material 3-Iodophenyl isocyanate
    Assay 97%

    GHS H Statement
    H331-H334-H302-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit