Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Iodopyridine, 99%

Click to view available options
Quantity:
5g
25g
1120-90-7
C5H4IN
204.998
MFCD00023553
XDELKSRGBLWMBA-UHFFFAOYSA-N
3-iodo pyridine,3-iodo-pyridine,pyridine, 3-iodo,3-iodpyridin,3-iodopyridine,pubchem6634,acmc-1bvap,ksc490s0t
70714
3-iodopyridine
C1=CC(=CN=C1)I
1120-90-7
C5H4IN
204.998
MFCD00023553
XDELKSRGBLWMBA-UHFFFAOYSA-N
3-iodo pyridine,3-iodo-pyridine,pyridine, 3-iodo,3-iodpyridin,3-iodopyridine,pubchem6634,acmc-1bvap,ksc490s0t
70714
3-iodopyridine
C1=CC(=CN=C1)I
Quantity 5g
Flash Point 106°C (222°F)
Melting Point 53°C to 56°C
Boiling Point 90°C to 92°C (14mmHg)
Beilstein 106059
Formula Weight 205
Percent Purity 99%
Chemical Name or Material 3-Iodopyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning