Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodopyridine 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
1120-90-7
C5H4IN
205
MFCD00023553
XDELKSRGBLWMBA-UHFFFAOYSA-N
3-iodo pyridine,3-iodo-pyridine,pyridine, 3-iodo,3-iodpyridin,3-iodopyridine,pubchem6634,acmc-1bvap,ksc490s0t
70714
3-iodopyridine
C1=CC(=CN=C1)I
1120-90-7
C5H4IN
205
MFCD00023553
XDELKSRGBLWMBA-UHFFFAOYSA-N
3-iodo pyridine,3-iodo-pyridine,pyridine, 3-iodo,3-iodpyridin,3-iodopyridine,pubchem6634,acmc-1bvap,ksc490s0t
70714
3-iodopyridine
C1=CC(=CN=C1)I
Flash Point 106°C
Melting Point 53.0°C to 56.0°C
Quantity 10g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 205
Percent Purity 99%
Chemical Name or Material 3-Iodopyridine, 99%

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-322-6

TSCA : TSCA