Learn More
Alfa Aesar™ 3-Isobutylpyridine, 99%
Specifications
Density | 0.909 |
Quantity | 10g |
Formula Weight | 135.21 |
Percent Purity | 99% |
Chemical Name or Material | 3-Isobutylpyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 238-003-6
TSCA : Yes
Recommended Storage : Ambient temperatures