Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Isopropoxycarbonyl)phenylboronic acid, 95%

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle
342002-80-6
C10H13BO4
208.02
HXVODQWMURGQMP-UHFFFAOYSA-N
3-isopropoxycarbonyl phenylboronic acid,3-isopropoxycarbonyl phenyl boronic acid,3-isopropoxycarbonyl benzeneboronic acid,3-isopropoxycarbonylphenylboronic acid,propan-2-yl 3-dihydroxyboranyl benzoate,3-isopropylcarbonyl phenylboronic acid,m-isopropoxycarbonyl phenylboronic acid,isopropyl 3-boronobenzoate,3-propan-2-yloxycarbonylphenyl boronic acid,acmc-209i5n
2734363
(3-propan-2-yloxycarbonylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)OC(C)C)(O)O
342002-80-6
C10H13BO4
208.02
HXVODQWMURGQMP-UHFFFAOYSA-N
3-isopropoxycarbonyl phenylboronic acid,3-isopropoxycarbonyl phenyl boronic acid,3-isopropoxycarbonyl benzeneboronic acid,3-isopropoxycarbonylphenylboronic acid,propan-2-yl 3-dihydroxyboranyl benzoate,3-isopropylcarbonyl phenylboronic acid,m-isopropoxycarbonyl phenylboronic acid,isopropyl 3-boronobenzoate,3-propan-2-yloxycarbonylphenyl boronic acid,acmc-209i5n
2734363
(3-propan-2-yloxycarbonylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)OC(C)C)(O)O
Quantity 250mg
Packaging Glass Bottle
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 208.02
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-(Isopropoxycarbonyl)phenylboronic acid
Show More Show Less