Alfa Aesar™ 3-Isopropoxypropylamine, 98+%

Manufacturer: Alfa Aesar™ B23748AD

Quantity 50mL

 View more versions of this product

Catalog No. AAB23748AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2906-12-9
    Molecular Formula (CH3)2CHO(CH2)3NH2
    Formula Weight 117.19
    MDL Number MFCD00008220
    UN Number UN2733
    Chemical Name or Material 3-Isopropoxypropylamine

    Specifications

    Formula Weight 117.19
    Quantity 50mL
    TSCA TSCA
    Molecular Formula (CH3)2CHO(CH2)3NH2
    MDL Number MFCD00008220
    CAS 2906-12-9
    UN Number UN2733
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Flash Point 42°C (108°F)
    Boiling Point 149° to 151°C
    Chemical Name or Material 3-Isopropoxypropylamine
    Assay ≥98%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit