Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Isopropoxypropylamine, 98+%

Click to view available options
Quantity:
50mL
250mL
2906-12-9
C6H15NO
117.19
MFCD00008220
VHYUNSUGCNKWSO-UHFFFAOYSA-N
3-isopropoxypropylamine,3-isopropoxy-1-propylamine,isopropoxypropylamine,1-propanamine, 3-1-methylethoxy,3-aminopropyl isopropyl ether,3-1-methylethoxy-1-propanamine,3-isopropoxy-n-propylamine,3-isopropoxypropan-1-amine,propylamine, 3-isopropoxy,3-propan-2-yloxy propan-1-amine
76194
3-propan-2-yloxypropan-1-amine
CC(C)OCCCN
2906-12-9
C6H15NO
117.19
MFCD00008220
VHYUNSUGCNKWSO-UHFFFAOYSA-N
3-isopropoxypropylamine,3-isopropoxy-1-propylamine,isopropoxypropylamine,1-propanamine, 3-1-methylethoxy,3-aminopropyl isopropyl ether,3-1-methylethoxy-1-propanamine,3-isopropoxy-n-propylamine,3-isopropoxypropan-1-amine,propylamine, 3-isopropoxy,3-propan-2-yloxy propan-1-amine
76194
3-propan-2-yloxypropan-1-amine
CC(C)OCCCN
Boiling Point 149°C to 151°C
Quantity 250mL
Flash Point 42°C (108°F)
UN Number UN2733
Formula Weight 117.19
Percent Purity ≥98%
Chemical Name or Material 3-Isopropoxypropylamine

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA