Alfa Aesar™ 3-Isopropyl-5-methylaniline hydrochloride, 98%

Manufacturer: Alfa Aesar™ H3385914

Quantity 25g

 View more versions of this product

Catalog No. AAH3385914

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C10H15N·HCl
    Formula Weight 185.7
    MDL Number MFCD22125030
    Chemical Name or Material 3-Isopropyl-5-methylaniline hydrochloride

    Specifications

    Formula Weight 185.7
    Molecular Formula C10H15N·HCl
    MDL Number MFCD22125030
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈225°C
    Quantity 25g
    Chemical Name or Material 3-Isopropyl-5-methylaniline hydrochloride
    Assay 98%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit