Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Acros Organics™

Click to view available options
Quantity:
100mg
1g
55750-62-4
C11H10N2O6
266.21
JKHVDAUOODACDU-UHFFFAOYSA-N
n-succinimidyl 3-maleimidopropionate,2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate,3-maleimido propionic acid n-hydroxysuccinimide ester,bmps,3-maleimidopropionic acid n-succinimidyl ester,3-maleimidopropionic acid nhs,succinimido 3-maleimidopropanoate,2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate,3-maleimidopropionic acid n-hydroxysuccinimide ester
4620597
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O
55750-62-4
C11H10N2O6
266.21
JKHVDAUOODACDU-UHFFFAOYSA-N
n-succinimidyl 3-maleimidopropionate,2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate,3-maleimido propionic acid n-hydroxysuccinimide ester,bmps,3-maleimidopropionic acid n-succinimidyl ester,3-maleimidopropionic acid nhs,succinimido 3-maleimidopropanoate,2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate,3-maleimidopropionic acid n-hydroxysuccinimide ester
4620597
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O
Packaging Glass Bottle
Infrared Spectrum Authentic
Melting Point 159.0°C to 160.0°C
Quantity 1g
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 266.21
Percent Purity 99%
Chemical Name or Material 3-Maleimidopropionic acid N-hydroxysuccinimide ester

GHS H Statement
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning