Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
96-27-5,7732-18-5
C3H8O2S
108.155
MFCD00004879
PJUIMOJAAPLTRJ-UHFFFAOYSA-N
3-mercapto-1,2-propanediol,monothioglycerol,1-thioglycerol,thioglycerol,thioglycerine,thioglycerin,thiovanol,3-mercaptopropane-1,2-diol,alpha-thioglycerol,glycerol-1-thiol
7291
CHEBI:74537
3-sulfanylpropane-1,2-diol
C(C(CS)O)O
96-27-5,7732-18-5
C3H8O2S
108.155
MFCD00004879
PJUIMOJAAPLTRJ-UHFFFAOYSA-N
3-mercapto-1,2-propanediol,monothioglycerol,1-thioglycerol,thioglycerol,thioglycerine,thioglycerin,thiovanol,3-mercaptopropane-1,2-diol,alpha-thioglycerol,glycerol-1-thiol
7291
CHEBI:74537
3-sulfanylpropane-1,2-diol
C(C(CS)O)O
Boiling Point 118.0°C (5.0 mmHg)
Color Colorless to Yellow
Density 1.2300g/mL
Specific Gravity 1.23
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25mL
Flash Point >112°C
Refractive Index 1.5073
CAS Min % 88.0,8.0
CAS Max % 92.0,12.0
Linear Formula HSCH2CH(OH)CH2OH
Formula Weight 108.16
Physical Form Viscous Solution
Percent Purity ≥88%
Chemical Name or Material 3-Mercapto-1, 2-propanediol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-495-0

RTECSNumber : TY8140000

TSCA : TSCA