Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Mercapto-1-hexanol, 96%

Click to view available options
Quantity:
5g
25g
51755-83-0
C6H14OS
134.237
MFCD00792515
TYZFMFVWHZKYSE-UHFFFAOYSA-N
3-mercapto-1-hexanol,3-mercaptohexanol,3-mercaptohexan-1-ol,1-hexanol, 3-mercapto,3-thiohexan-1-ol,3-sulphanylhexan-1-ol,3-sulfanyl-1-hexanol,3-thiohexanol,acmc-209kvm,3-mercapto-hexan-1-ol
521348
CHEBI:77690
3-sulfanylhexan-1-ol
CCCC(CCO)S
51755-83-0
C6H14OS
134.237
MFCD00792515
TYZFMFVWHZKYSE-UHFFFAOYSA-N
3-mercapto-1-hexanol,3-mercaptohexanol,3-mercaptohexan-1-ol,1-hexanol, 3-mercapto,3-thiohexan-1-ol,3-sulphanylhexan-1-ol,3-sulfanyl-1-hexanol,3-thiohexanol,acmc-209kvm,3-mercapto-hexan-1-ol
521348
CHEBI:77690
3-sulfanylhexan-1-ol
CCCC(CCO)S
Applications
3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Boiling Point 250°C
Flash Point >100°C (212°F)
Quantity 5g
Beilstein 1846873
Formula Weight 134.24
Percent Purity 96%
Chemical Name or Material 3-Mercapto-1-hexanol

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA