Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-1-propanol, 97+%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
19721-22-3
C3H8OS
92.16
MFCD00192262
SHLSSLVZXJBVHE-UHFFFAOYSA-N
3-mercapto-1-propanol,1-propanol, 3-mercapto,3-mercaptopropanol,3-hydroxypropanethiol,3-hydroxypropyl mercaptan,3-mecapto-1-propanol,3-mecaptopropan-1-ol,1-mercapto-3-propanol,3-hydroxypropylmercaptan,3-mercaptopropan-1-ol
88211
3-sulfanylpropan-1-ol
C(CO)CS
19721-22-3
C3H8OS
92.16
MFCD00192262
SHLSSLVZXJBVHE-UHFFFAOYSA-N
3-mercapto-1-propanol,1-propanol, 3-mercapto,3-mercaptopropanol,3-hydroxypropanethiol,3-hydroxypropyl mercaptan,3-mecapto-1-propanol,3-mecaptopropan-1-ol,1-mercapto-3-propanol,3-hydroxypropylmercaptan,3-mercaptopropan-1-ol
88211
3-sulfanylpropan-1-ol
C(CO)CS
Boiling Point 106°C (50.0mmHg)
Flash Point 85°C
Specific Gravity 1.062
Density 1.0620g/mL
Refractive Index 1.4950 to 1.497
Color Colorless
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97% min. (GC)
Linear Formula SH(CH2)3OH
Solubility Information Solubility in water: slightly soluble
Formula Weight 92.16
Physical Form Liquid
Percent Purity 97+%
Chemical Name or Material 3-Mercapto-1-propanol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger