Alfa Aesar™ (3-Mercaptopropyl)methyldimethoxysilane, 95%

Manufacturer: Alfa Aesar™ 42426K2

Quantity 1L

 View more versions of this product

Catalog No. AA42426K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31001-77-1
    Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
    Formula Weight 180.34
    MDL Number MFCD00053923
    Synonym 3-(Dimethoxymethylsilyl)-1-propanethiol
    Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane

    Specifications

    Formula Weight 180.34
    Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
    Synonym 3-(Dimethoxymethylsilyl)-1-propanethiol
    Color Colorless
    Quantity 1L
    Flash Point 93°C (199°F)
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Density 1.000g/mL
    TSCA TSCA
    Physical Form Liquid
    Sensitivity Moisture Sensitive
    MDL Number MFCD00053923
    Boiling Point 96°C (30mmHg)
    Health Hazard 1 Warning
    Refractive Index 1.4500
    CAS 31001-77-1
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
    Assay 95%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit