Learn More
3-(Methacryloyloxy)propyltrimethoxysilane, 97%, Thermo Scientific Chemicals
Description
3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
Solubility
Miscible with water.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Boiling Point | 190°C |
Quantity | 100 g |
Beilstein | 1952435 |
Flash Point | 92°C (197°F) |
Solubility Information | Miscible with water. |
SMILES | CC(=C)C(=O)OCCC[Si](OC)(OC)OC |
Formula Weight | 248.35 |
Percent Purity | 97% |
Refractive Index | 1.431 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 219-785-8 |
RTECSNumber | UC0230000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.