Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methanesulfonyl-2-oxo-1-imidazolidinecarbonyl Chloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
41762-76-9
C5H7ClN2O4S
226.631
MFCD00272360
ZWTPALHHEULAPI-UHFFFAOYSA-N
1-Chlorocarbonyl-3-methanesulfonyl-2-imidazolidinone, 1-Chloroformyl-3-methanesulfonyl-2-imidazolidinone
3016299
3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl chloride
CS(=O)(=O)N1CCN(C1=O)C(=O)Cl
41762-76-9
C5H7ClN2O4S
226.631
MFCD00272360
ZWTPALHHEULAPI-UHFFFAOYSA-N
1-Chlorocarbonyl-3-methanesulfonyl-2-imidazolidinone, 1-Chloroformyl-3-methanesulfonyl-2-imidazolidinone
3016299
3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl chloride
CS(=O)(=O)N1CCN(C1=O)C(=O)Cl
Color White-Yellow
Melting Point 180°C
UN Number 3261
Quantity 5g
Formula Weight 226.63
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 3-Methanesulfonyl-2-oxo-1-imidazolidinecarbonyl Chloride