Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-1,2-propanediol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
623-39-2
C4H10O3
106.12
MFCD00004719
PSJBSUHYCGQTHZ-UHFFFAOYSA-N
3-methoxy-1,2-propanediol,1,2-propanediol, 3-methoxy,1-o-methylglycerol,glycerol 1-monomethyl ether,glycerin-alpha-monomethyl ether,glycerin-.alpha.-monomethyl ether,glycerin methyl ether,3-methoxypropan-1,2-diol,1-o-methyl-rac-glycerol,acmc-1bhm7
92822
3-methoxypropane-1,2-diol
COCC(CO)O
623-39-2
C4H10O3
106.12
MFCD00004719
PSJBSUHYCGQTHZ-UHFFFAOYSA-N
3-methoxy-1,2-propanediol,1,2-propanediol, 3-methoxy,1-o-methylglycerol,glycerol 1-monomethyl ether,glycerin-alpha-monomethyl ether,glycerin-.alpha.-monomethyl ether,glycerin methyl ether,3-methoxypropan-1,2-diol,1-o-methyl-rac-glycerol,acmc-1bhm7
92822
3-methoxypropane-1,2-diol
COCC(CO)O
Refractive Index 1.4430 to 1.445
Density 1.1200g/mL
Boiling Point 133°C (35.0mmHg)
Flash Point >110°C
Specific Gravity 1.12
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula H3COCH2CH(OH)CH2OH
Solubility Information Solubility in water: soluble in water
Formula Weight 106.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methoxy-1, 2-propanediol
Show More Show Less

EINECSNumber : 210-791-6