Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-1-propanol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1589-49-7
C4H10O2
90.12
JDFDHBSESGTDAL-UHFFFAOYSA-N
3-methoxy-1-propanol,1-propanol, 3-methoxy,3-methoxypropanol,trimethylene glycol monomethyl ether,beta-pgme,1,3-propanediol monomethyl ether,unii-oak281wz1p,propylene glycol monomethyl ether, beta,oak281wz1p,mpol
74116
3-methoxypropan-1-ol
COCCCO
1589-49-7
C4H10O2
90.12
JDFDHBSESGTDAL-UHFFFAOYSA-N
3-methoxy-1-propanol,1-propanol, 3-methoxy,3-methoxypropanol,trimethylene glycol monomethyl ether,beta-pgme,1,3-propanediol monomethyl ether,unii-oak281wz1p,propylene glycol monomethyl ether, beta,oak281wz1p,mpol
74116
3-methoxypropan-1-ol
COCCCO
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Specific Gravity 0.942
Color Colorless
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Density 0.9420g/mL
Flash Point 38°C
Boiling Point 149.0°C to 150.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3O(CH2)3OH
Formula Weight 90.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methoxy-1-propanol
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning