Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-2-methylpyridine, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
26395-26-6
C7H9NO
123.16
QRCUQLANPHYVEH-UHFFFAOYSA-N
3-methoxy-2-picoline,3-methoxy-2-methyl-pyridine,pyridine,3-methoxy-2-methyl,pubchem6637,2-methyl-3-methoxypyridine
3015270
3-methoxy-2-methylpyridine
CC1=C(C=CC=N1)OC
26395-26-6
C7H9NO
123.16
QRCUQLANPHYVEH-UHFFFAOYSA-N
3-methoxy-2-picoline,3-methoxy-2-methyl-pyridine,pyridine,3-methoxy-2-methyl,pubchem6637,2-methyl-3-methoxypyridine
3015270
3-methoxy-2-methylpyridine
CC1=C(C=CC=N1)OC
Packaging Glass Bottle
Infrared Spectrum Authentic
Flash Point 61°C
Boiling Point 170.0°C to 175.0°C
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 123.16
Percent Purity 97%
Chemical Name or Material 3-Methoxy-2-methylpyridine

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 247-652-4