Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-2-nitrobenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
53055-05-3
C8H7NO4
181.15
MFCD00007135
GDTUACILWWLIJF-UHFFFAOYSA-N
benzaldehyde, 3-methoxy-2-nitro,3-methoxy-2-nitro-benzaldehyde,2-nitro-3-methoxybenzaldehyde,pubchem13093,acmc-20agz4,2-nitro-3-methoxy benzaldehyde,benzaldehyde,3-methoxy-2-nitro,3-methoxy-2-nitrobenzaldehyde
98867
3-methoxy-2-nitrobenzaldehyde
COC1=CC=CC(=C1[N+](=O)[O-])C=O
53055-05-3
C8H7NO4
181.15
MFCD00007135
GDTUACILWWLIJF-UHFFFAOYSA-N
benzaldehyde, 3-methoxy-2-nitro,3-methoxy-2-nitro-benzaldehyde,2-nitro-3-methoxybenzaldehyde,pubchem13093,acmc-20agz4,2-nitro-3-methoxy benzaldehyde,benzaldehyde,3-methoxy-2-nitro,3-methoxy-2-nitrobenzaldehyde
98867
3-methoxy-2-nitrobenzaldehyde
COC1=CC=CC(=C1[N+](=O)[O-])C=O
Color Beige to White or Yellow
Melting Point 97.0°C to 101.0°C
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3OC6H3(NO2)CHO
Beilstein 08, 62
Formula Weight 181.15
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Methoxy-2-nitrobenzaldehyde
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-332-9