Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-4-methylbenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
7151-68-0
C9H10O3
166.176
MFCD00002566
CEAVPXDEPGAVDA-UHFFFAOYSA-N
4-methyl-m-anisic acid,3-methoxy-p-toluic acid,benzoic acid, 3-methoxy-4-methyl,3-methoxy-4-methyl-benzoic acid,3-methoxy-4-methyl benzoic acid,3-methoxy-p-toluic acid; 4-methyl-m-anisic acid,3-methoxy-4-methylbenzoicacid,pubchem2672,5-carboxy-2-methylanisole,acmc-209oj0
81579
3-methoxy-4-methylbenzoic acid
CC1=C(C=C(C=C1)C(=O)O)OC
7151-68-0
C9H10O3
166.176
MFCD00002566
CEAVPXDEPGAVDA-UHFFFAOYSA-N
4-methyl-m-anisic acid,3-methoxy-p-toluic acid,benzoic acid, 3-methoxy-4-methyl,3-methoxy-4-methyl-benzoic acid,3-methoxy-4-methyl benzoic acid,3-methoxy-p-toluic acid; 4-methyl-m-anisic acid,3-methoxy-4-methylbenzoicacid,pubchem2672,5-carboxy-2-methylanisole,acmc-209oj0
81579
3-methoxy-4-methylbenzoic acid
CC1=C(C=C(C=C1)C(=O)O)OC
Quantity 5g
Melting Point 154°C to 160°C
Color Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H3(CH3)CO2H
Beilstein 10, 238
Formula Weight 166.18
Physical Form Fine Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Methoxy-4-methylbenzoic acid, 97%
Show More Show Less