Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methoxy-4-methylbenzoic acid, 99%

Manufacturer:  Alfa Aesar™ B2298203

Quantity 1g

 View more versions of this product

Catalog No. AAB2298203


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7151-68-0
166.18
1938389
3-Methoxy-4-methylbenzoic acid
C9H10O3
MFCD00002566
3-Methoxy-p-toluic acid; 4-Methyl-m-anisic acid

Specifications

158° to 160°C
7151-68-0
166.18
1938389
3-Methoxy-4-methylbenzoic acid
1g
C9H10O3
MFCD00002566
3-Methoxy-p-toluic acid; 4-Methyl-m-anisic acid
99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit