Alfa Aesar™ 3-Methoxy-4-methylbenzoic acid, 99%

Manufacturer: Alfa Aesar™ B2298214

Quantity 25g

 View more versions of this product

Catalog No. AAB2298214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7151-68-0
    Molecular Formula C9H10O3
    Formula Weight 166.18
    MDL Number MFCD00002566
    Beilstein 1938389
    Synonym 3-Methoxy-p-toluic acid; 4-Methyl-m-anisic acid
    Chemical Name or Material 3-Methoxy-4-methylbenzoic acid

    Specifications

    Formula Weight 166.18
    Molecular Formula C9H10O3
    MDL Number MFCD00002566
    CAS 7151-68-0
    Beilstein 1938389
    Synonym 3-Methoxy-p-toluic acid; 4-Methyl-m-anisic acid
    Melting Point 158° to 160°C
    Quantity 25g
    Chemical Name or Material 3-Methoxy-4-methylbenzoic acid
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit