Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-4-nitrobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
5081-36-7
C8H7NO5
197.146
MFCD00007353
PWURRRRGLCVBMX-UHFFFAOYSA-N
3-methoxy-4-nitrobenzoic acid,4-nitro-3-methoxybenzoic acid,4-nitro-m-anisic acid,benzoic acid, 3-methoxy-4-nitro,3-methoxy-4-nitrobenzoicacid,pubchem3918,acmc-1bn9s,ksc270m6t,3-methoxy4-nitro-benzoic acid,3-methoxy-4-nitro-benzoic acid
78764
COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]
5081-36-7
C8H7NO5
197.146
MFCD00007353
PWURRRRGLCVBMX-UHFFFAOYSA-N
3-methoxy-4-nitrobenzoic acid,4-nitro-3-methoxybenzoic acid,4-nitro-m-anisic acid,benzoic acid, 3-methoxy-4-nitro,3-methoxy-4-nitrobenzoicacid,pubchem3918,acmc-1bn9s,ksc270m6t,3-methoxy4-nitro-benzoic acid,3-methoxy-4-nitro-benzoic acid
78764
COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]
Quantity 10g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H3(NO2)CO2H
Beilstein 10, 146
Formula Weight 197.15
Chemical Name or Material 3-Methoxy-4-nitrobenzoic acid, 98%