Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Methoxyacetophenone, 98%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
586-37-8
C9H10O2
150.18
MFCD00008736
BAYUSCHCCGXLAY-UHFFFAOYSA-N
3'-methoxyacetophenone,3-methoxyacetophenone,3-acetylanisole,m-methoxyacetophenone,1-3-methoxyphenyl ethanone,ethanone, 1-3-methoxyphenyl,m-acetanisole,acetophenone, m-methoxy,1-3-methoxyphenyl ethan-1-one,acetophenone, 3'-methoxy
11460
1-(3-methoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OC
586-37-8
C9H10O2
150.18
MFCD00008736
BAYUSCHCCGXLAY-UHFFFAOYSA-N
3'-methoxyacetophenone,3-methoxyacetophenone,3-acetylanisole,m-methoxyacetophenone,1-3-methoxyphenyl ethanone,ethanone, 1-3-methoxyphenyl,m-acetanisole,acetophenone, m-methoxy,1-3-methoxyphenyl ethan-1-one,acetophenone, 3'-methoxy
11460
1-(3-methoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OC
Refractive Index 1.5394 to 1.5414
Packaging Glass bottle
Flash Point 110°C
Specific Gravity 1.09
Density 1.0900g/mL
Quantity 50mL
Boiling Point 239.0°C to 241.0°C
Color Brown to Yellow
Melting Point -7.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3OC6H4COCH3
Beilstein 08, 86
Solubility Information Solubility in water: immiscible
Formula Weight 150.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3'-Methoxyacetophenone, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-573-3

TSCA : TSCA