Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Methoxyacetophenone 98%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
586-37-8
C9H10O2
150.18
MFCD00008736
BAYUSCHCCGXLAY-UHFFFAOYSA-N
3'-methoxyacetophenone,3-methoxyacetophenone,3-acetylanisole,m-methoxyacetophenone,1-3-methoxyphenyl ethanone,ethanone, 1-3-methoxyphenyl,m-acetanisole,acetophenone, m-methoxy,1-3-methoxyphenyl ethan-1-one,acetophenone, 3'-methoxy
11460
1-(3-methoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OC
586-37-8
C9H10O2
150.18
MFCD00008736
BAYUSCHCCGXLAY-UHFFFAOYSA-N
3'-methoxyacetophenone,3-methoxyacetophenone,3-acetylanisole,m-methoxyacetophenone,1-3-methoxyphenyl ethanone,ethanone, 1-3-methoxyphenyl,m-acetanisole,acetophenone, m-methoxy,1-3-methoxyphenyl ethan-1-one,acetophenone, 3'-methoxy
11460
1-(3-methoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OC
Melting Point -7.0°C
Refractive Index 1.5394 to 1.5414
Specific Gravity 1.09
Packaging Glass bottle
Flash Point 110°C
Quantity 250mL
Infrared Spectrum Authentic
Color Brown to Yellow
Density 1.0900g/mL
Boiling Point 239.0°C to 241.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4COCH3
Beilstein 08, 86
Solubility Information Solubility in water: immiscible
Formula Weight 150.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3'-Methoxyacetophenone, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 209-573-3

TSCA : TSCA