Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methoxybenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
10365-98-7
C7H9BO3
151.956
MFCD00161359
NLLGFYPSWCMUIV-UHFFFAOYSA-N
3-methoxyphenyl boronic acid,3-methoxybenzeneboronic acid,3-methoxyphenyl boranediol,3-methoxypenylboronic acid,3-boronanisole,3-boronoanisole,boronic acid, 3-methoxyphenyl,m-methoxyphenylboronic acid,3-methoxy phenylboronic acid
2734370
(3-methoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OC)(O)O
10365-98-7
C7H9BO3
151.956
MFCD00161359
NLLGFYPSWCMUIV-UHFFFAOYSA-N
3-methoxyphenyl boronic acid,3-methoxybenzeneboronic acid,3-methoxyphenyl boranediol,3-methoxypenylboronic acid,3-boronanisole,3-boronoanisole,boronic acid, 3-methoxyphenyl,m-methoxyphenylboronic acid,3-methoxy phenylboronic acid
2734370
(3-methoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OC)(O)O
Melting Point 158°C to 162°C
Quantity 5g
Beilstein 2831223
Formula Weight 151.96
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning