Alfa Aesar™ 3-Methoxybenzoic acid, 99%

Manufacturer: Alfa Aesar™ A107670B

Quantity 1000g

 View more versions of this product

Catalog No. AAA107670B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 586-38-9
    Molecular Formula C8H8O3
    Formula Weight 152.15
    MDL Number MFCD00002499
    Beilstein 508838
    Synonym m-Anisic acid
    Chemical Name or Material 3-Methoxybenzoic acid

    Specifications

    Formula Weight 152.15
    TSCA TSCA
    Synonym m-Anisic acid
    Boiling Point 170° to 172°C (10mmHg)
    Molecular Formula C8H8O3
    CAS 586-38-9
    Beilstein 508838
    RTECS Number BZ4375000
    Odor Odorless
    Melting Point 104° to 108°C
    MDL Number MFCD00002499
    Quantity 1000g
    Chemical Name or Material 3-Methoxybenzoic acid
    Assay 99%

    Description

    Applications
    3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

    Solubility
    Soluble in 95% ethanol (50 mg/ml), water (2 mg/ml at 25°C) and methanol.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit