Alfa Aesar™ 3-Methoxybenzonitrile, 98%

Manufacturer: Alfa Aesar™ B2417106

Quantity 5g

 View more versions of this product

Catalog No. AAB2417106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1527-89-5
    Molecular Formula C8H7NO
    Formula Weight 133.15
    MDL Number MFCD00001801
    UN Number UN3439
    Beilstein 1932666
    Synonym m-Anisonitrile
    Chemical Name or Material 3-Methoxybenzonitrile

    Specifications

    Formula Weight 133.15
    Melting Point 22° to 24°C
    Molecular Formula C8H7NO
    MDL Number MFCD00001801
    CAS 1527-89-5
    UN Number UN3439
    Synonym m-Anisonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 105°C (221°F)
    Odor Almond-like
    Boiling Point 110° to 111°C (12mmHg)
    Quantity 5g
    Beilstein 1932666
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 3-Methoxybenzonitrile
    Assay 98%

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit