Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
1527-89-5
C8H7NO
133.15
MFCD00001801
KLXSUMLEPNAZFK-UHFFFAOYSA-N
m-cyanoanisole,benzonitrile, 3-methoxy,m-anisonitrile,m-methoxybenzonitrile,3-cyanoanisole,3-anisonitrile,benzonitrile, m-methoxy,3-methoxy-benzonitrile,3-methoxy benzonitrile,m-methoxybenzontrile
73712
3-methoxybenzonitrile
COC1=CC=CC(=C1)C#N
1527-89-5
C8H7NO
133.15
MFCD00001801
KLXSUMLEPNAZFK-UHFFFAOYSA-N
m-cyanoanisole,benzonitrile, 3-methoxy,m-anisonitrile,m-methoxybenzonitrile,3-cyanoanisole,3-anisonitrile,benzonitrile, m-methoxy,3-methoxy-benzonitrile,3-methoxy benzonitrile,m-methoxybenzontrile
73712
3-methoxybenzonitrile
COC1=CC=CC(=C1)C#N
Refractive Index 1.5392 to 1.5412 (20°C, 589nm)
Quantity 25mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.089g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4CN
Beilstein 10, II, 82
Formula Weight 133.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3-Methoxybenzonitrile, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-201-3

TSCA : TSCA