Alfa Aesar™ 3-Methoxybenzoyl chloride, 99%

Manufacturer: Alfa Aesar™ B2357109

Quantity 10g

 View more versions of this product

Catalog No. AAB2357109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1711-05-3
    Molecular Formula C8H7ClO2
    Formula Weight 170.6
    MDL Number MFCD00000673
    UN Number UN1729
    Beilstein 386659
    Synonym m-Anisoyl chloride
    Chemical Name or Material 3-Methoxybenzoyl chloride

    Specifications

    Formula Weight 170.6
    Flash Point 92°C (197°F)
    Quantity 10g
    Molecular Formula C8H7ClO2
    MDL Number MFCD00000673
    CAS 1711-05-3
    UN Number UN1729
    Beilstein 386659
    Synonym m-Anisoyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANISOYL CHLORIDE
    Boiling Point 123° to 125°C (15mmHg)
    Chemical Name or Material 3-Methoxybenzoyl chloride
    Assay 99%

    Description

    Solubility
    Soluble in chloroform. Reacts with water.

    Notes
    Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit