Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybenzyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
824-98-6
C8H9ClO
156.61
MFCD00000907
VGISFWWEOGVMED-UHFFFAOYSA-N
3-methoxybenzyl chloride,1-chloromethyl-3-methoxybenzene,m-methoxybenzyl chloride,m-chloromethyl anisole,benzene, 1-chloromethyl-3-methoxy,anisole, m-chloromethyl,3-methoxybenzylchloride,3-chloromethyl anisole,3-methoxy benzyl chloride,benzyl chloride,3-methoxy
69994
1-(chloromethyl)-3-methoxybenzene
COC1=CC=CC(=C1)CCl
824-98-6
C8H9ClO
156.61
MFCD00000907
VGISFWWEOGVMED-UHFFFAOYSA-N
3-methoxybenzyl chloride,1-chloromethyl-3-methoxybenzene,m-methoxybenzyl chloride,m-chloromethyl anisole,benzene, 1-chloromethyl-3-methoxy,anisole, m-chloromethyl,3-methoxybenzylchloride,3-chloromethyl anisole,3-methoxy benzyl chloride,benzyl chloride,3-methoxy
69994
1-(chloromethyl)-3-methoxybenzene
COC1=CC=CC(=C1)CCl
Specific Gravity 1.07
Refractive Index 1.5430 to 1.545
Flash Point 101°C
Boiling Point 124°C (13.0mmHg)
Density 1.0700g/mL
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3OC6H4CH2Cl
Beilstein 06, I, 189
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzyl chloride, 97%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 212-541-1

TSCA : TSCA